Learn More
Selleck Chemical LLC Homatropine Bromide S4025-50mg
SDP
Catalog No. 503109648
Encompass_Preferred
Homatropine Bromide is an muscarinic AChR antagonist inhibits endothelial and smooth muscle muscarinic receptors of WKY-E and SHR-E with IC50 of 162 5 nM and 170 3 nM respectively
Catalog No. 50-310-9648 Supplier Selleck Chemical LLC Supplier No. S402550MG
Homatropine Bromide is an muscarinic AChR antagonist inhibits endothelial and smooth muscle muscarinic receptors of WKY-E and SHR-E with IC50 of 162 5 nM and 170 3 nM respectively