Learn More
Apexbio Technology LLC Homatropine Bromide 51-56-9 5g
SDP
Catalog No. 5000089837
Encompass_Preferred
Homatropine Bromide (CAS 51-56-9) is a small molecule antagonist of muscarinic acetylcholine receptors (AChRs) It selectively inhibits muscarinic receptor-mediated responses in endothelial and smooth muscle cells with reported IC50 values of 162 5 nM and 170 3 nM for WKY-E and SHR-E cell models respectively This compound serves as a valuable tool for investigating the role of muscarinic signaling in vascular function and smooth muscle physiology within experimental research settings
Catalog No. 50-000-89837 Supplier Apexbio Technology LLC Supplier No. B16035000
Homatropine Bromide (CAS 51-56-9) is a small molecule antagonist of muscarinic acetylcholine receptors (AChRs) It selectively inhibits muscarinic receptor-mediated responses in endothelial and smooth muscle cells with reported IC50 values of 162 5 nM and 170 3 nM for WKY-E and SHR-E cell models respectively This compound serves as a valuable tool for investigating the role of muscarinic signaling in vascular function and smooth muscle physiology within experimental research settings