Learn More
Selleck Chemical LLC Homatropine Methylbromide S4024-1g
SDP
Catalog No. 503108838
Encompass_Preferred
Homatropine Methylbromide is muscarinic AChR antagonist inhibits endothelial and smooth muscle muscarinic receptors of WKY-E and SHR-E with IC50 of 162 5 nM and 170 3 nM respectively
Catalog No. 50-310-8838 Supplier Selleck Chemical LLC Supplier No. S40241G
Homatropine Methylbromide is muscarinic AChR antagonist inhibits endothelial and smooth muscle muscarinic receptors of WKY-E and SHR-E with IC50 of 162 5 nM and 170 3 nM respectively