Learn More
Apexbio Technology LLC Homatropine Methylbromide 80-49-9 10mM (in 1mL DMSO)
SDP
Catalog No. 5000089838
Encompass_Preferred
Homatropine Methylbromide (CAS 80-49-9) is a small molecule antagonist of muscarinic acetylcholine receptors (AChRs) It inhibits muscarinic signaling by blocking receptor activity on endothelial and smooth muscle cells as demonstrated in WKY-E and SHR-E models with IC50 values of 162 5 nM and 170 3 nM respectively This compound is utilized in studies investigating cholinergic modulation of vascular tone and smooth muscle function offering a tool for dissecting the roles of muscarinic receptors in cardiovascular and autonomic research contexts
Catalog No. 50-000-89838 Supplier Apexbio Technology LLC Supplier No. B16045.1
Homatropine Methylbromide (CAS 80-49-9) is a small molecule antagonist of muscarinic acetylcholine receptors (AChRs) It inhibits muscarinic signaling by blocking receptor activity on endothelial and smooth muscle cells as demonstrated in WKY-E and SHR-E models with IC50 values of 162 5 nM and 170 3 nM respectively This compound is utilized in studies investigating cholinergic modulation of vascular tone and smooth muscle function offering a tool for dissecting the roles of muscarinic receptors in cardiovascular and autonomic research contexts