Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Homatropine methylbromide | 80-49-9 | 98.0% | 10 MM * 1 ML
SDP

Catalog No. 5000413077
Encompass_Preferred

Homatropine methylbromide is a muscarinic AChR antagonist. It inhibits muscarinic receptors in both endothelial and smooth muscle, with IC50 values of 162.5 nM and 170.3 nM in WKY-E and SHR-E, respectively. It has been shown to prevent lethality in a rat model of acute organophosphate poisoning.

  • Muscarinic AChR antagonist
  • Inhibits endothelial and smooth muscle muscarinic receptors
  • IC50 values of 162.5 nM (WKY-E) and 170.3 nM (SHR-E)
  • Produces a dose ratio of 259 in atrium from guinea-pigs (20 μM)
  • Comparable to atropine in preventing lethality in a rat model of acute organophosphate poisoning (20 mg/kg)
  • Molecular Weight: 370.28
  • Formula: C17H24BrNO3
  • Appearance: Solid, white to off-white

Catalog No. 50-004-13077 Supplier Medchemexpress LLC Supplier No. HYB138810MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Homatropine methylbromide is a muscarinic AChR antagonist. It inhibits muscarinic receptors in both endothelial and smooth muscle, with IC50 values of 162.5 nM and 170.3 nM in WKY-E and SHR-E, respectively. It has been shown to prevent lethality in a rat model of acute organophosphate poisoning.

  • Muscarinic AChR antagonist
  • Inhibits endothelial and smooth muscle muscarinic receptors
  • IC50 values of 162.5 nM (WKY-E) and 170.3 nM (SHR-E)
  • Produces a dose ratio of 259 in atrium from guinea-pigs (20 μM)
  • Comparable to atropine in preventing lethality in a rat model of acute organophosphate poisoning (20 mg/kg)
  • Molecular Weight: 370.28
  • Formula: C17H24BrNO3
  • Appearance: Solid, white to off-white

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.