Learn More
Aviva Systems Biology Corp HOMER1 Antibody - N-terminal region (ARP40182_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40182_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HOMER1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HOMER1. Alternate names: HOMER1A, HOMER1B, HOMER1C, SYN47, Ves-1, HOMERApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57325