Learn More
Aviva Systems Biology Corp HOMER3 Antibody - N-terminal region (ARP62178_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62178_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HOMER3. It was validated on Western Blot. . Alternate names: HOMER-3, VESL3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-287-601