Learn More
Aviva Systems Biology Corp HOMEZ Antibody - N-terminal region (ARP37601_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37601_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against HOMEZ. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of mouse HOMEZ. Alternate names: mKIAA1443ApplicationWB. Species reactivity: Mouse, Rat
Catalog No. 50-014-53082