Learn More
Medchemexpress LLC HY-111594 5mg Medchemexpress, Homo-PROTAC cereblon degrader 1 CAS:2244520-98-5 Purity:>98%
SDP
Catalog No. 501932377
Encompass_Preferred
Medchemexpress, HY-111594 5mg Homo-PROTAC cereblon degrader 1 CAS:2244520-98-5 Homo-PROTAC cereblon degrader 1 (compound 15a) is a highly potent and efficient cereblon (CRBN) degrader with only minimal effects on IKZF1 and IKZF3 [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2377 Supplier Medchemexpress LLC Supplier No. HY1115945MG
Medchemexpress, HY-111594 5mg Homo-PROTAC cereblon degrader 1 CAS:2244520-98-5 Homo-PROTAC cereblon degrader 1 (compound 15a) is a highly potent and efficient cereblon (CRBN) degrader with only minimal effects on IKZF1 and IKZF3 [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.