Learn More
Medchemexpress LLC HY-111593 5mg Medchemexpress, Homo-PROTAC pVHL30 degrader 1 CAS:2244684-49-7 Purity:>98%
SDP
Catalog No. 501932376
Encompass_Preferred
Medchemexpress, HY-111593 5mg Homo-PROTAC pVHL30 degrader 1 CAS:2244684-49-7 Homo-PROTAC pVHL30 degrader 1 is a potent pVHL30 degrader based on PROTAC [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2376 Supplier Medchemexpress LLC Supplier No. HY1115935MG
Medchemexpress, HY-111593 5mg Homo-PROTAC pVHL30 degrader 1 CAS:2244684-49-7 Homo-PROTAC pVHL30 degrader 1 is a potent pVHL30 degrader based on PROTAC [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.