Learn More
Medchemexpress LLC HY-109564 10mg Medchemexpress, 5α-Pregnane-3β,6α-diol-20-one CAS:21853-11-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10956410MG
Encompass_Preferred
Medchemexpress, HY-111593 10mg Homo-PROTAC pVHL30 degrader 1 CAS:2244684-49-7 Homo-PROTAC pVHL30 degrader 1 is a potent pVHL30 degrader based on PROTAC. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8706