Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Homo sildenafil | 642928-07-2 | 99.3% | C23H32N6O4S | 10MG
SDP

Catalog No. 5000261896
Encompass_Preferred

Homo sildenafil is a small-molecule analog of sildenafil that functions as a phosphodiesterase (PDE) inhibitor. It is supplied as a solid research compound intended for biochemical and pharmacological studies.

  • Acts as a phosphodiesterase inhibitor.
  • Molecular formula: C23H32N6O4S.
  • Molecular weight: 488.6 Da.
  • Purity: 99.3%.
  • Supplied as a solid for research use.
  • Available in multiple package sizes for assay flexibility.
  • Suitable for biochemical and pharmacological research applications.

Catalog No. 50-002-61896 Supplier Medchemexpress LLC Supplier No. HY13110010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Homo sildenafil is a small-molecule analog of sildenafil that functions as a phosphodiesterase (PDE) inhibitor. It is supplied as a solid research compound intended for biochemical and pharmacological studies.

  • Acts as a phosphodiesterase inhibitor.
  • Molecular formula: C23H32N6O4S.
  • Molecular weight: 488.6 Da.
  • Purity: 99.3%.
  • Supplied as a solid for research use.
  • Available in multiple package sizes for assay flexibility.
  • Suitable for biochemical and pharmacological research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.