Learn More
Medchemexpress LLC Homo sildenafil | 642928-07-2 | MFCD09752148 | 99.3% | 488.60 | C23H32N6O4S | 5 MG
SDP
Catalog No. 5000263985
Encompass_Preferred
Homo sildenafil is a small-molecule analog of sildenafil that functions as a phosphodiesterase (PDE) inhibitor for research and analytical applications. The compound is supplied as a solid with molecular formula C23H32N6O4S and molecular weight 488.60, and is accompanied by analytical documentation for quality verification.
  • Acts as a phosphodiesterase inhibitor for biochemical and pharmacological studies.
  • High purity (99.3%), suitable for analytical applications.
  • Provided as a solid for convenient handling and storage.
  • Molecular formula C23H32N6O4S and molecular weight 488.60.
  • Analytical documents available: COA, SDS, HNMR, LCMS.
Catalog No. 50-002-63985 Supplier Medchemexpress LLC Supplier No. HY1311005MG
Homo sildenafil is a small-molecule analog of sildenafil that functions as a phosphodiesterase (PDE) inhibitor for research and analytical applications. The compound is supplied as a solid with molecular formula C23H32N6O4S and molecular weight 488.60, and is accompanied by analytical documentation for quality verification.
  • Acts as a phosphodiesterase inhibitor for biochemical and pharmacological studies.
  • High purity (99.3%), suitable for analytical applications.
  • Provided as a solid for convenient handling and storage.
  • Molecular formula C23H32N6O4S and molecular weight 488.60.
  • Analytical documents available: COA, SDS, HNMR, LCMS.