Learn More
Cayman Chemical Homobrassinolide, Purity: ≥85%, CAS: 74174-44-0, 100 mg
SDP
Catalog No. 502911658
Encompass_Preferred
A brassinosteroid; field application increases the number of fruits per plant, fruit length, and fruit breadth in tomato plants at 0.12 g/ha
Catalog No. 50-291-1658 Supplier Cayman Chemical Supplier No. 36038100 MG
A brassinosteroid; field application increases the number of fruits per plant, fruit length, and fruit breadth in tomato plants at 0.12 g/ha