Learn More
Cayman Chemical HomobrasInolIde 10mg
SDP
Catalog No. 502911664
Encompass_Preferred
A brassinosteroid; field application increases the number of fruits per plant, fruit length, and fruit breadth in tomato plants at 0.12 g/ha
Catalog No. 50-291-1664 Supplier Cayman Chemical Supplier No. 3603810 MG
A brassinosteroid; field application increases the number of fruits per plant, fruit length, and fruit breadth in tomato plants at 0.12 g/ha