Learn More
Cayman Chemical Homobrassinolide, Purity: ≥85%, CAS: 74174-44-0, 50 mg
SDP
Catalog No. 502912816
Encompass_Preferred
A brassinosteroid; field application increases the number of fruits per plant, fruit length, and fruit breadth in tomato plants at 0.12 g/ha
Catalog No. 50-291-2816 Supplier Cayman Chemical Supplier No. 3603850 MG
A brassinosteroid; field application increases the number of fruits per plant, fruit length, and fruit breadth in tomato plants at 0.12 g/ha