Learn More
Medchemexpress LLC HY-117389 5mg Medchemexpress, Homocarbonyltopsentin CAS:172286-77-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1173895MG
Encompass_Preferred
Medchemexpress, HY-117389 5mg Homocarbonyltopsentin CAS:172286-77-0 Homocarbonyltopsentin (PK4C9) is a small-molecule TSL2-binding compound, binds to pentaloop conformations of TSL2 and promotes a shift to triloop conformations that display enhanced SMN2 exon 7 (E7) splicing with EC50 value of 16 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9471