Learn More
Cayman Chemical Homocarnosine, Purity: ≥95%, CAS: 3650-73-5, 10 mg
SDP
Catalog No. 502950566
Encompass_Preferred
A dipeptide; inhibits AAPH-induced lipid peroxidation at 10 mM; decreases LDH and PGE2 release from PC12 cells in an in vitro model of ischemia induced by oxygen-glucose deprivation at 1 mM; reduces infarct area and neurological deficit scores in a rat model of MCAO-induced cerebral ischemia-reperfusion injury; reduces mortality in a mouse model of subcutaneous S. aureus infection at 5 mg/animal
Catalog No. 50-295-0566 Supplier Cayman Chemical Supplier No. 3369510 MG
A dipeptide; inhibits AAPH-induced lipid peroxidation at 10 mM; decreases LDH and PGE2 release from PC12 cells in an in vitro model of ischemia induced by oxygen-glucose deprivation at 1 mM; reduces infarct area and neurological deficit scores in a rat model of MCAO-induced cerebral ischemia-reperfusion injury; reduces mortality in a mouse model of subcutaneous S. aureus infection at 5 mg/animal