Learn More
Cayman Chemical Homocitrulline, Purity: ≥95%, CAS: 1190-49-4, 100 mg
SDP
Catalog No. 502910835
Encompass_Preferred
An amino acid and a product of carbamylation; protein-bound homocitrulline levels are increased in the aortas of transgenic Ldlr-/- mice expressing human MPO fed a high-fat atherogenic diet; homocitrulline-containing peptides induce the development of arthritis in mice; homocitrulline-containing peptide and anti-homocitrulline antibody levels are increased in whole blood from patients with erosive rheumatoid arthritis compared to patients with non-erosive rheumatoid arthritis
Catalog No. 50-291-0835 Supplier Cayman Chemical Supplier No. 34980100 MG
An amino acid and a product of carbamylation; protein-bound homocitrulline levels are increased in the aortas of transgenic Ldlr-/- mice expressing human MPO fed a high-fat atherogenic diet; homocitrulline-containing peptides induce the development of arthritis in mice; homocitrulline-containing peptide and anti-homocitrulline antibody levels are increased in whole blood from patients with erosive rheumatoid arthritis compared to patients with non-erosive rheumatoid arthritis