Learn More
Cayman Chemical HomocItrulIn-d3 1mg
SDP
Supplier:  Cayman Chemical 363391 MG
Encompass_Preferred
An internal standard for the quantification of homocitrulline by GC- or LC-MS
Catalog No. 50-291-2107