Learn More
Cayman Chemical HomocItrulIn-d3 500ug
SDP
Catalog No. 502912348
Encompass_Preferred
An internal standard for the quantification of homocitrulline by GC- or LC-MS
Catalog No. 50-291-2348 Supplier Cayman Chemical Supplier No. 36339500 UG
An internal standard for the quantification of homocitrulline by GC- or LC-MS