Learn More
Cusabio Technology LLC Homocysteic acid Hcy
Catalog No. 5000049061
Encompass_Preferred
This kit detects Hcy in rat serum plasma and tissue homogenates using a quantitative sandwich ELISA method It offers a detection range of 0 78 nmol/mL to 50 nmol/mL with a sensitivity of 0 195 nmol/mL ensuring precise measurement of low-abundance samples The assay requires a short incubation time of 1-5 hours and utilizes a sample volume of 50-100 L per test Results are measured at an absorbance wavelength of 450 nm providing reliable and reproducible data for research applications Designed for flexibility this kit is suitable for analyzing various biological matrices to study homocysteic acid levels in experimental models
Catalog No. 50-000-49061 Supplier Cusabio Technology LLC Supplier No. CSBE08896R
This kit detects Hcy in rat serum plasma and tissue homogenates using a quantitative sandwich ELISA method It offers a detection range of 0 78 nmol/mL to 50 nmol/mL with a sensitivity of 0 195 nmol/mL ensuring precise measurement of low-abundance samples The assay requires a short incubation time of 1-5 hours and utilizes a sample volume of 50-100 L per test Results are measured at an absorbance wavelength of 450 nm providing reliable and reproducible data for research applications Designed for flexibility this kit is suitable for analyzing various biological matrices to study homocysteic acid levels in experimental models