Learn More
Creative Biomart Becaplermin, Recombinant Human PDGFB
Catalog No. 502084235
Encompass_Preferred
The product is produced by recombinant DNA technology by insertion of the gene for the B chain of platelet derived growth factor (PDGF) into the yeast, Saccharomyces cerevisiae. The product has a molecular weight of approximately 25 KD and is a homodimer composed of two identical polypeptide chains that are bound together by disulfide bonds.
Catalog No. 50-208-4235 Supplier Creative Biomart Supplier No. THP0184
The product is produced by recombinant DNA technology by insertion of the gene for the B chain of platelet derived growth factor (PDGF) into the yeast, Saccharomyces cerevisiae. The product has a molecular weight of approximately 25 KD and is a homodimer composed of two identical polypeptide chains that are bound together by disulfide bonds.