Learn More
Chemglass Life Sciences Homogenizer, Bio-Gen, 110V
SDP
Catalog No. 501311366
Encompass
Homogenizer, Bio-Gen, 110V. Homogenizers are ideal for plant and animal homogenization, PCR applications, soil and wastewater extractions, and Karl Fischer titration. These high-speed rotor/stator homogenizers feature infinitely variable speed control and can be used to rapidly crush, tear, disintegrate, emulsify, or suspend particles in a liquid medium. The systems impart shear energy into a liquid emulsion or suspension far greater than that of a conventional mixer and its able to homogenize samples with volumes as small as 0.03mL or as large as 1L*. The accessory dispersing elements provide a wide range of volumes and applications. Available in 110 or 220 volt. (*Higher volumes dependent on viscosity.)
Catalog No. 50-131-1366 Supplier Chemglass Life Sciences Supplier No. CLS503015
Homogenizer, Bio-Gen, 110V. Homogenizers are ideal for plant and animal homogenization, PCR applications, soil and wastewater extractions, and Karl Fischer titration. These high-speed rotor/stator homogenizers feature infinitely variable speed control and can be used to rapidly crush, tear, disintegrate, emulsify, or suspend particles in a liquid medium. The systems impart shear energy into a liquid emulsion or suspension far greater than that of a conventional mixer and its able to homogenize samples with volumes as small as 0.03mL or as large as 1L*. The accessory dispersing elements provide a wide range of volumes and applications. Available in 110 or 220 volt. (*Higher volumes dependent on viscosity.)