Learn More
Bellco Glass, Inc. Homogenizer Drive,115V
SDP
Catalog No. 501945185
Encompass_Preferred
DOES NOT COME WITH SUPPORT STAND!!! STAND IS #1981-30000!!!Description: Homogenizer Drive, 115V. Homogenizer drives are used as a source of motor for disaggregating tissue samples into individual cells. The drive is a motor that can be set to a special Rpm depending upon the type of cells being used. The drives are typically bolted to an upright stand for support during the operation. Our stand is a general purpose one, however, any lab stand would be appropriate. Customers may already have either a support stand or a "flex-a-frame" lab framework that is used to attach many types of laboratory glassware/equipment.PERMANENT MAGNET MOTOR, 5/16"OD SHAFTSHIGH SPEED SHAFT; 4000rpm (26 in-lb TORQUE)LOW SPEED SHAFT; 333rpm (3.6 in-lb TORQUE)ELECTRONIC MOTOR CONTROLLERSWITCH FOR REVERSING PESTLE DIRECTION
Catalog No. 50-194-5185 Supplier Bellco Glass, Inc. Supplier No. 198120115
DOES NOT COME WITH SUPPORT STAND!!! STAND IS #1981-30000!!!Description: Homogenizer Drive, 115V. Homogenizer drives are used as a source of motor for disaggregating tissue samples into individual cells. The drive is a motor that can be set to a special Rpm depending upon the type of cells being used. The drives are typically bolted to an upright stand for support during the operation. Our stand is a general purpose one, however, any lab stand would be appropriate. Customers may already have either a support stand or a "flex-a-frame" lab framework that is used to attach many types of laboratory glassware/equipment.PERMANENT MAGNET MOTOR, 5/16"OD SHAFTSHIGH SPEED SHAFT; 4000rpm (26 in-lb TORQUE)LOW SPEED SHAFT; 333rpm (3.6 in-lb TORQUE)ELECTRONIC MOTOR CONTROLLERSWITCH FOR REVERSING PESTLE DIRECTION