Learn More
Glas Col LLC Homogenizer
SDP
Catalog No. NC1100042
Encompass_Preferred
Stirrer motor homogenizer and speed controller for gentle dispersion of tissue and cellsAnalog unit with a variable speed of 333 to 4,000rpm 120V and an electronic motor controller that maintains speed regardless of changes in viscosity1/4hp motor drive unit has a geared output shaft for higher torque applications and lower speeds, and a direct-drive output shaft for lower torque applications and higher speedsElectronic motor controller provides smooth rotation and repeatability while reducing motor temperature and noiseReversing switch on controller allows motor shaft to rotate the pestle clockwise or counterclockwise during homogenization
Catalog No. NC1100042 Supplier Glas Col LLC Supplier No. 099C K54
Stirrer motor homogenizer and speed controller for gentle dispersion of tissue and cellsAnalog unit with a variable speed of 333 to 4,000rpm 120V and an electronic motor controller that maintains speed regardless of changes in viscosity1/4hp motor drive unit has a geared output shaft for higher torque applications and lower speeds, and a direct-drive output shaft for lower torque applications and higher speedsElectronic motor controller provides smooth rotation and repeatability while reducing motor temperature and noiseReversing switch on controller allows motor shaft to rotate the pestle clockwise or counterclockwise during homogenization