Learn More
Medchemexpress LLC Homogentisic acid | 451-13-8 | MFCD00004324 | 99.5% | 168.15 g·mol⁻¹ | C8H8O4 | 5 MG
SDP
Catalog No. 5000264242
Encompass_Preferred
Homogentisic acid analytical standard is a reference-grade phenolic metabolite used for research and analytical applications, including method development and quantitation by HPLC, GC, and MS. It supports diagnostic and research workflows for detecting homogentisic acid, a metabolite that accumulates in urine and serum, and is supplied as a stable solid with recommended storage conditions to preserve integrity.
  • High purity (99.5%) suitable for analytical references.
  • Suitable for HPLC, GC, and MS method development and quantitation.
  • Applicable to diagnostic studies of urine and serum metabolites.
  • Supplied as a white to off-white solid for convenient handling.
  • Stable when stored under recommended conditions to maintain longevity.
Catalog No. 50-002-64242 Supplier Medchemexpress LLC Supplier No. HY113283R5MG
Add to Cart
Add to Cart
Homogentisic acid analytical standard is a reference-grade phenolic metabolite used for research and analytical applications, including method development and quantitation by HPLC, GC, and MS. It supports diagnostic and research workflows for detecting homogentisic acid, a metabolite that accumulates in urine and serum, and is supplied as a stable solid with recommended storage conditions to preserve integrity.
  • High purity (99.5%) suitable for analytical references.
  • Suitable for HPLC, GC, and MS method development and quantitation.
  • Applicable to diagnostic studies of urine and serum metabolites.
  • Supplied as a white to off-white solid for convenient handling.
  • Stable when stored under recommended conditions to maintain longevity.