Learn More
Cayman Chemical Homogentisic Acid, Purity: ≥95%, CAS: 451-13-8, 50 mg
SDP
Catalog No. 502984020
Encompass_Preferred
An intermediate in the catabolism of aromatic amino acids; formed in excess during alkaptonuria
Catalog No. 50-298-4020 Supplier Cayman Chemical Supplier No. 2004550 MG
An intermediate in the catabolism of aromatic amino acids; formed in excess during alkaptonuria