Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Homogentisic acid, ≥98% (GC), MilliporeSigma™ Supelco™

Catalog No. 111002099 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-2099 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-2099 Supplier MilliporeSigma Supelco Supplier No. 9216825MG

missing translation for 'validMainframeMsg'

Featured Industries: Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

CAS 451-13-8
Linear Formula (HO)2C6H3 CH2 CO2 H
MDL Number MFCD00004324
Melting Point 150°C to 152°C
Synonym 2,5-Dihydroxyphenylacetic acid
Type Analytical Standard
Quantity 25 mg
Formula Weight 168.15
Percent Purity ≥98% (GC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.