Learn More
Cayman Chemical Homogentisic Acid lactone, Purity: ≥95%, CAS: 2688-48-4, 500 mg
SDP
Catalog No. 502943766
Encompass_Preferred
A fungal metabolite with fungicidal activity; active against the plant pathogenic fungus F. graminearum; has been used as a substrate in the determination of PON1 activity; has been used in the chemiluminescent detection of HRP release from liposomes; an intermediate in the synthesis of the near-infrared lipid droplet probe LD-2
Catalog No. 50-294-3766 Supplier Cayman Chemical Supplier No. 41254500 MG
A fungal metabolite with fungicidal activity; active against the plant pathogenic fungus F. graminearum; has been used as a substrate in the determination of PON1 activity; has been used in the chemiluminescent detection of HRP release from liposomes; an intermediate in the synthesis of the near-infrared lipid droplet probe LD-2