Learn More
Medchemexpress LLC Homogentisic acid lactone | 2688-48-4 | MFCD00005857 | 99.1% | 150.13 g/mol | C8H6O3 | 250 MG
SDP
Catalog No. 5000302371
Encompass_Preferred
Homogentisic acid lactone is a fungal metabolite supplied as a research reagent. It exhibits fungicidal activity and is used as a substrate for measuring serum paraoxonase/arylesterase 1 (PON1) activity in biochemical assays.
  • Shows fungicidal activity against Fusarium graminearum.
  • Can be used as a substrate for serum paraoxonase/arylesterase 1 (PON1) activity assays.
  • High purity suitable for analytical and biochemical applications.
  • Well-characterized molecular properties (C8H6O3, 150.13 g/mol).
  • Supplied as a research-grade reagent for laboratory studies.
Catalog No. 50-003-02371 Supplier Medchemexpress LLC Supplier No. 250MGHYW017445
Homogentisic acid lactone is a fungal metabolite supplied as a research reagent. It exhibits fungicidal activity and is used as a substrate for measuring serum paraoxonase/arylesterase 1 (PON1) activity in biochemical assays.
  • Shows fungicidal activity against Fusarium graminearum.
  • Can be used as a substrate for serum paraoxonase/arylesterase 1 (PON1) activity assays.
  • High purity suitable for analytical and biochemical applications.
  • Well-characterized molecular properties (C8H6O3, 150.13 g/mol).
  • Supplied as a research-grade reagent for laboratory studies.