Learn More
Medchemexpress LLC HY-14944 50mg Medchemexpress, Homoharringtonine CAS:26833-87-4 Purity:>98%
SDP
Catalog No. 502029077
Encompass_Preferred
Medchemexpress, HY-14944 50mg Homoharringtonine CAS:26833-87-4 Homoharringtonine (Omacetaxine mepesuccinate;HHT) is a cytotoxic alkaloid with antitumor properties which acts by inhibiting translation elongation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9077 Supplier Medchemexpress LLC Supplier No. HY1494450MG
Medchemexpress, HY-14944 50mg Homoharringtonine CAS:26833-87-4 Homoharringtonine (Omacetaxine mepesuccinate;HHT) is a cytotoxic alkaloid with antitumor properties which acts by inhibiting translation elongation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.