Learn More
Enzo Life Sciences Homoharringtonine (5mg). CAS: 26833-87-4
SDP
Supplier:  Enzo Life Sciences ALX350236M005
Encompass_Preferred
Cephalotaxine alkaloid. Inhibits the formation of diphenylalanine and acetylphenylalanyl-puromycin in liver ribosomes. Has promising activity in hematologic malignancies. Lowers the levels of telomerase. Induces apoptosis. Purity: ≥96% (Assay). Solubility: Soluble in DMSO or 100% ethanol. Long Term Storage: +4°C.
Catalog No. 50-200-8792