Learn More
Cell Signaling Technology Homologous Recombination DNA Repair Antibody Sampler Kit, Unconjugated, 1 Kit
SDP
Catalog No. 502051371
Encompass
The Homologous Recombination (HR) DNA Repair Antibody Sampler Kit provides an economical means for the detection of proteins involved in HR DNA repair pathways. Each antibody is included in a quantity sufficient to perform two western blot experiments.
  • Includes nine primary antibodies for monitoring DNA repair factors
  • Contains an HRP-linked secondary antibody for signal detection
  • Validated for use in western blotting applications
  • Provided in a stable storage buffer containing glycerol and sodium azide
  • Designed for research use only
Catalog No. 50-205-1371 Supplier Cell Signaling Technology Supplier No. 99891T
The Homologous Recombination (HR) DNA Repair Antibody Sampler Kit provides an economical means for the detection of proteins involved in HR DNA repair pathways. Each antibody is included in a quantity sufficient to perform two western blot experiments.
  • Includes nine primary antibodies for monitoring DNA repair factors
  • Contains an HRP-linked secondary antibody for signal detection
  • Validated for use in western blotting applications
  • Provided in a stable storage buffer containing glycerol and sodium azide
  • Designed for research use only