Learn More
Medchemexpress LLC HY-N1949 5mg Medchemexpress, Homoplantaginin CAS:17680-84-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN19495MG
Encompass_Preferred
Medchemexpress, HY-N1949 5mg Homoplantaginin CAS:17680-84-1 Homoplantaginin is a flavonoid from a traditional Chinese medicine Salvia plebeia with antiinflammatory and antioxidant properties. Homoplantaginin could inhibit TNF-α and IL-6 mRNA expression, IKKβ and NF-κB phosphorylation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3864