Learn More
Medchemexpress LLC Homosalate | 118-56-9 | 99.4% | 10 G
SDP
Catalog No. 5000408481
Encompass_Preferred
Homosalate (Homomenthyl salicylate) is an organic compound utilized as a sunscreen to filter UV rays and shield the skin from sun damage. It also possesses anti-inflammatory properties.
  • Filters UV rays and shields skin from sun damage
  • Possesses anti-inflammatory properties
  • Exhibits cytotoxic and genotoxic effects on MCF-7 cells
  • Promotes release of tumor-derived extracellular vesicles
  • Inhibits HTR8/Svneo cell invasion
Catalog No. 50-004-08481 Supplier Medchemexpress LLC Supplier No. HYB092810G
Add to Cart
Add to Cart
Homosalate (Homomenthyl salicylate) is an organic compound utilized as a sunscreen to filter UV rays and shield the skin from sun damage. It also possesses anti-inflammatory properties.
  • Filters UV rays and shields skin from sun damage
  • Possesses anti-inflammatory properties
  • Exhibits cytotoxic and genotoxic effects on MCF-7 cells
  • Promotes release of tumor-derived extracellular vesicles
  • Inhibits HTR8/Svneo cell invasion