Learn More
Medchemexpress LLC Homosalate-d4 (homomenthyl salicylate-d4) | 118-56-9 | 99.9% | 1 MG
SDP
Catalog No. 5000409293
Encompass_Preferred
Homosalate-d4 is the deuterium labeled version of Homosalate, an organic compound used in sunscreens as a chemical UV filter, protecting the skin from sun damage.
  • Can be used as a tracer for quantitation during drug development.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-09293 Supplier Medchemexpress LLC Supplier No. HYB0928S1MG
Homosalate-d4 is the deuterium labeled version of Homosalate, an organic compound used in sunscreens as a chemical UV filter, protecting the skin from sun damage.
  • Can be used as a tracer for quantitation during drug development.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.