Learn More
Medchemexpress LLC Homosalate-d4 (Homomenthyl salicylate-d4) | 118-56-9 | 99.95% | 5 MG
SDP
Catalog No. 5000408637
Encompass_Preferred
Homosalate-d4 is the deuterium labeled Homosalate, an organic compound used in sunscreens as a chemical UV filter to protect the skin from sun damage. Intended for research use only, this compound can serve as a tracer and an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes, like deuterium, are incorporated into drug molecules for quantitation during drug development, with deuteration showing potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-08637 Supplier Medchemexpress LLC Supplier No. HYB0928S5MG
Homosalate-d4 is the deuterium labeled Homosalate, an organic compound used in sunscreens as a chemical UV filter to protect the skin from sun damage. Intended for research use only, this compound can serve as a tracer and an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes, like deuterium, are incorporated into drug molecules for quantitation during drug development, with deuteration showing potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS