Learn More
Cayman Chemical N-(-ketocaproyl)-L-Homoserine lactone, 143537-62-6, 50 mg
SDP
Supplier:  Cayman Chemical 1001120750
Encompass_Preferred
Molecular Formula C10H15NO4, Purity ≥95%, Formula Weight 213.2, Synonyms: 3-O-C6-(L)-HSL; A HSL compound produced in the marine bioluminescence bacterium V. fischeri.
Catalog No. NC0371586