Learn More
Medchemexpress LLC HY-N0384 100mg Medchemexpress, Homovanillic acid CAS:306-08-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN0384100MG
Encompass_Preferred
Medchemexpress, HY-N0384 100mg Homovanillic acid CAS:306-08-1 Homovanillic acid is a dopamine metabolite found to be associated with aromatic L-amino acid decarboxylase deficiency, celiac disease, growth hormone deficiency, and sepiapterin reductase deficiency. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0263