Learn More
Medchemexpress LLC Homovanillic acid | 306-08-1 | 182.17 | 1 G
SDP
Catalog No. 5000390058
Encompass_Preferred
Homovanillic acid (Vanilacetic acid) is a dopamine metabolite found to be associated with aromatic L-amino acid decarboxylase deficiency, celiac disease, growth hormone deficiency, and sepiapterin reductase deficiency. This compound offers valuable insights for various research applications.
  • Increases presynaptic membrane protein SYN1 in HT22 and rat hippocampal neurons.
  • Protects synaptic structure.
  • Inhibits the degradation of autophagy marker proteins LC3 and SQSTM1/p62.
  • Reduces the formation of autophagosomes.
  • Exhibits antidepressant activities in mouse Corticosterone-induced depression models.
  • Exhibits neuroprotective activities in mouse Corticosterone-induced depression models.
Catalog No. 50-003-90058 Supplier Medchemexpress LLC Supplier No. HYN03841G
Homovanillic acid (Vanilacetic acid) is a dopamine metabolite found to be associated with aromatic L-amino acid decarboxylase deficiency, celiac disease, growth hormone deficiency, and sepiapterin reductase deficiency. This compound offers valuable insights for various research applications.
  • Increases presynaptic membrane protein SYN1 in HT22 and rat hippocampal neurons.
  • Protects synaptic structure.
  • Inhibits the degradation of autophagy marker proteins LC3 and SQSTM1/p62.
  • Reduces the formation of autophagosomes.
  • Exhibits antidepressant activities in mouse Corticosterone-induced depression models.
  • Exhibits neuroprotective activities in mouse Corticosterone-induced depression models.