Learn More
Medchemexpress LLC Homovanillic acid | 306-08-1 | 182.17 | 50 MG
SDP
Catalog No. 5000390085
Encompass_Preferred
Homovanillic acid (Standard) is the analytical standard of Homovanillic acid (HY-N0384). This product is intended for research and analytical applications. Homovanillic acid (Vanilacetic acid) is a dopamine metabolite found to be associated with aromatic L-amino acid decarboxylase deficiency, celiac disease, growth hormone deficiency, and sepiapterin reductase deficiency.
  • Used as a reference standard.
  • Utilized in qualitative, quantitative, and methodological research experiments.
  • Applied in HPLC, GC, and MS analyses.
Catalog No. 50-003-90085 Supplier Medchemexpress LLC Supplier No. HYN0384R50MG
Homovanillic acid (Standard) is the analytical standard of Homovanillic acid (HY-N0384). This product is intended for research and analytical applications. Homovanillic acid (Vanilacetic acid) is a dopamine metabolite found to be associated with aromatic L-amino acid decarboxylase deficiency, celiac disease, growth hormone deficiency, and sepiapterin reductase deficiency.
  • Used as a reference standard.
  • Utilized in qualitative, quantitative, and methodological research experiments.
  • Applied in HPLC, GC, and MS analyses.