Learn More
Medchemexpress LLC Homovanillic acid | 306-08-1 | MFCD00004350 | 98.1% | 182.17 | C9H10O4 | 10 MG
SDP
Catalog No. 5000296065
Encompass_Preferred
Homovanillic acid analytical standard supplied as a 10 mg reference material for research and analytical applications. It is a dopamine metabolite commonly used as a calibration standard in biochemical and clinical assays; key properties include molecular formula C9H10O4, molecular weight 182.17 g/mol, CAS 306-08-1, and high purity for quantitative work.
  • Analytical standard suitable for HPLC and mass spectrometry.
  • 10 mg package for small-scale assays and calibration.
  • High purity (98.1%) for reliable calibration and quantitation.
  • Molecular weight 182.17 g/mol; molecular formula C9H10O4.
  • Used as a reference in studies of dopamine metabolism and related assays.
Catalog No. 50-002-96065 Supplier Medchemexpress LLC Supplier No. HYN0384R10MG
Homovanillic acid analytical standard supplied as a 10 mg reference material for research and analytical applications. It is a dopamine metabolite commonly used as a calibration standard in biochemical and clinical assays; key properties include molecular formula C9H10O4, molecular weight 182.17 g/mol, CAS 306-08-1, and high purity for quantitative work.
  • Analytical standard suitable for HPLC and mass spectrometry.
  • 10 mg package for small-scale assays and calibration.
  • High purity (98.1%) for reliable calibration and quantitation.
  • Molecular weight 182.17 g/mol; molecular formula C9H10O4.
  • Used as a reference in studies of dopamine metabolism and related assays.