Learn More
Medchemexpress LLC Homovanillic acid-d5 | 53587-32-9 | 187.20 | 5 MG
SDP
Catalog No. 5000389040
Encompass_Preferred
Homovanillic acid-d5 (Vanilacetic acid-d5) is the deuterium labeled Homovanillic acid. This compound is a dopamine metabolite found to be associated with aromatic L-amino acid decarboxylase deficiency, celiac disease, growth hormone deficiency, and sepiapterin reductase deficiency.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-89040 Supplier Medchemexpress LLC Supplier No. HYN0384S25MG
Homovanillic acid-d5 (Vanilacetic acid-d5) is the deuterium labeled Homovanillic acid. This compound is a dopamine metabolite found to be associated with aromatic L-amino acid decarboxylase deficiency, celiac disease, growth hormone deficiency, and sepiapterin reductase deficiency.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.