Learn More
Cusabio Technology LLC Homovanillic Acid HVA
Catalog No. 5000052126
Encompass_Preferred
This Rat HVA ELISA Kit was designed for the quantitative measurement of Rat HVA protein in serum plasma tissue homogenates It is a Competitive ELISA kit its detection range is 0 049 ng/mL-200 ng/mL and the sensitivity is 0 049 ng/mL
Catalog No. 50-000-52126 Supplier Cusabio Technology LLC Supplier No. CSBEQ027785RA
This Rat HVA ELISA Kit was designed for the quantitative measurement of Rat HVA protein in serum plasma tissue homogenates It is a Competitive ELISA kit its detection range is 0 049 ng/mL-200 ng/mL and the sensitivity is 0 049 ng/mL