Learn More
Medchemexpress LLC Homovanillic acid (Standard) | 306-08-1 | 182.17 | 100 MG
SDP
Catalog No. 5000389335
Encompass_Preferred
Homovanillic acid (Standard) (Vanilacetic acid (Standard)) is the analytical standard of Homovanillic acid (HY-N0384). This product is intended for research and analytical applications. Homovanillic acid (Vanilacetic acid) is a dopamine metabolite found to be associated with aromatic L-amino acid decarboxylase deficiency, celiac disease, growth hormone deficiency, and sepiapterin reductase deficiency.
  • Used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
  • Analytical standard grade.
  • Reference standard supplied assay.
Catalog No. 50-003-89335 Supplier Medchemexpress LLC Supplier No. HYN0384R100MG
Homovanillic acid (Standard) (Vanilacetic acid (Standard)) is the analytical standard of Homovanillic acid (HY-N0384). This product is intended for research and analytical applications. Homovanillic acid (Vanilacetic acid) is a dopamine metabolite found to be associated with aromatic L-amino acid decarboxylase deficiency, celiac disease, growth hormone deficiency, and sepiapterin reductase deficiency.
  • Used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
  • Analytical standard grade.
  • Reference standard supplied assay.