Learn More
Medchemexpress LLC Homovanillic acid-13C6,18O | 202468-52-8 | 190.13 | 1 MG
SDP
Catalog No. 5000388837
Encompass_Preferred
Homovanillic acid-13C6,18O is a 13C- and 18O-labeled homovanillic acid, a dopamine metabolite. It has been associated with conditions such as aromatic L-amino acid decarboxylase deficiency, celiac disease, growth hormone deficiency, and sepiapterin reductase deficiency. This compound is intended for research use only.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-88837 Supplier Medchemexpress LLC Supplier No. HYN0384S31MG
Homovanillic acid-13C6,18O is a 13C- and 18O-labeled homovanillic acid, a dopamine metabolite. It has been associated with conditions such as aromatic L-amino acid decarboxylase deficiency, celiac disease, growth hormone deficiency, and sepiapterin reductase deficiency. This compound is intended for research use only.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS