Learn More
Medchemexpress LLC Homovanillyl alcohol | 2380-78-1 | 100 MG
SDP
Catalog No. 5000417668
Encompass_Preferred
Homovanillyl alcohol is a biological metabolite of Hydroxytyrosol, a phenolic compound present in virgin olive oil (VOO) and wine. It protects red blood cells (RBCs) from oxidative injury and has a protective effect on cardiovascular disease. This product is intended for research use only.
  • Biological metabolite of Hydroxytyrosol
  • Found in virgin olive oil and wine
  • Protects red blood cells from oxidative injury
  • Exhibits a protective effect against cardiovascular disease
  • Intended for research use only
Catalog No. 50-004-17668 Supplier Medchemexpress LLC Supplier No. HYN7513100MG
Homovanillyl alcohol is a biological metabolite of Hydroxytyrosol, a phenolic compound present in virgin olive oil (VOO) and wine. It protects red blood cells (RBCs) from oxidative injury and has a protective effect on cardiovascular disease. This product is intended for research use only.
  • Biological metabolite of Hydroxytyrosol
  • Found in virgin olive oil and wine
  • Protects red blood cells from oxidative injury
  • Exhibits a protective effect against cardiovascular disease
  • Intended for research use only