Learn More
Cayman Chemical Homovanillic Acid, Purity: ≥98%, CAS: 306-08-1, 500 mg
SDP
Catalog No. 502940905
Encompass_Preferred
A dopamine metabolite; formed via deamination of dopamine by MAO to produce DOPAC followed by DOPAC metabolism by COMT; undergoes hydrogen peroxide-dependent oxidation in the presence of horseradish peroxidase to form a fluorescent dimer with ex/em = 312/420 nm, respectively; has been used to quantify hydrogen peroxide production in macrophages and neutrophils
Catalog No. 50-294-0905 Supplier Cayman Chemical Supplier No. 27307500 MG
A dopamine metabolite; formed via deamination of dopamine by MAO to produce DOPAC followed by DOPAC metabolism by COMT; undergoes hydrogen peroxide-dependent oxidation in the presence of horseradish peroxidase to form a fluorescent dimer with ex/em = 312/420 nm, respectively; has been used to quantify hydrogen peroxide production in macrophages and neutrophils