Learn More
Cayman Chemical Homovanillic Acid sulfate (sodium salt), Purity: ≥98%, 10 mg
SDP
Catalog No. 502983144
Encompass_Preferred
A phenol found in olive oil and a dopamine metabolite; a component of LDL in humans that consume olive oil; levels are increased in response to acute renal injury in humans and rats; has been used as a biomarker for early detection of kidney disease in pediatric nephrology
Catalog No. 50-298-3144 Supplier Cayman Chemical Supplier No. 2087710 MG
A phenol found in olive oil and a dopamine metabolite; a component of LDL in humans that consume olive oil; levels are increased in response to acute renal injury in humans and rats; has been used as a biomarker for early detection of kidney disease in pediatric nephrology